Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Spain
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Recombinant Human SHH (from E. coli)
Recombinant Human SHH (from E. coli)
Recombinant Human SHH (from E. coli)
Catalog # ABNOP9080.25UG
Proveedor:
Abnova
Número CAS:
Expand image
Recombinant Human SHH (from E. coli)
Catalog # ABNOP9080.25UG
Proveedor:
Abnova
Número CAS:
Especificaciones
Pk:
25 µG
Protein/peptide type:
Recombinant
Source:
E. coli
Species:
Human
Gene ID:
Q15465
Protein/peptide name:
SHH
Especificaciones
Acerca de este artículo
Product Details
Product Details