Recombinant Human CXCL8 (from E. coli), His-tag
Catalog # ABNOP9484.50UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Environmentally Preferable:
- Tag sequence:His-tag
- Gene ID:P10145
- Protein/peptide name:CXCL8
- Sequence:23a.a.-99a.a.