Order Entry
Spain
ContactUsLinkComponent
 
Recombinant Mouse CTLA4 (from Baculovirus), hIgG-His-tag
  ABNOP8533.20UG
 :  Abnova
 :  
Recombinant Mouse CTLA4 (from Baculovirus), hIgG-His-tag
  ABNOP8533.20UG
 :  Abnova
 :  

 

  • Pk:
    20 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Baculovirus
  • Species:
    Mouse
  • Environmentally Preferable:
  • Tag sequence:
    hIgG-His-tag
  • Gene ID:
    P16410
  • Protein/peptide name:
    CTLA4
  • Sequence:
    36a.a.-161a.a