Order Entry
Spain
ContactUsLinkComponent
Recombinant IL2 (from E. coli), His-tag
Recombinant IL2 (from E. coli), His-tag
Catálogo # ABNOP8254.5UG
Supplier:  Abnova
CAS Number:  
Recombinant IL2 (from E. coli), His-tag
Catálogo # ABNOP8254.5UG
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    5 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Tag sequence:
    His-tag
  • Gene ID:
    Q07885
  • Protein/peptide name:
    IL2
  • Sequence:
    21a.a.-154a.a.

Specifications

About this item