Order Entry
Spain
ContactUsLinkComponent
Recombinant Human FAS (from E. coli), His-tag
Recombinant Human FAS (from E. coli), His-tag
Catalog # ABNOP8581.20UG
Supplier:  Abnova
CAS Number:  
Recombinant Human FAS (from E. coli), His-tag
Catalog # ABNOP8581.20UG
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    20 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    His-tag
  • Protein/peptide name:
    FAS

Specifications

About this item