Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Spain
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Recombinant Human FGF16 (from E. coli)
Recombinant Human FGF16 (from E. coli)
Recombinant Human FGF16 (from E. coli)
Catalog # ABNOP8615.25UG
Supplier:
Abnova
CAS Number:
Ampliar imagen
Recombinant Human FGF16 (from E. coli)
Catalog # ABNOP8615.25UG
Supplier:
Abnova
CAS Number:
Specifications
Pk:
25 µG
Protein/peptide type:
Recombinant
Source:
E. coli
Species:
Human
Protein/peptide name:
FGF16
Specifications
About this item
Detalles del producto
Detalles del producto