Recombinant Mouse IGFBP6 (from Sf9 cells), His-tag
Catalog # ABNOP8199.10UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Source:Insect cells (Sf9)
- Species:Mouse
- Tag sequence:His-tag
- Gene ID:P47880
- Protein/peptide name:IGFBP6
- Sequence:26a.a.-238a.a.