Order Entry
Spain
ContactUsLinkComponent
Recombinant Mouse RSPO1 (from Sf9 cells), hIgG-His-tag
Recombinant Mouse RSPO1 (from Sf9 cells), hIgG-His-tag
Catalog # ABNOP8780.10UG
Supplier:  Abnova
CAS Number:  
Recombinant Mouse RSPO1 (from Sf9 cells), hIgG-His-tag
Catalog # ABNOP8780.10UG
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Insect cells (Sf9)
  • Species:
    Mouse
  • Tag sequence:
    hIgG-His-tag
  • Gene ID:
    Q9Z132
  • Protein/peptide name:
    RSPO1
  • Sequence:
    18a.a.-286a.a

Specifications

About this item