Recombinant Mouse BLMH (from E. coli), His-tag
Catalog # ABNOP8059.50UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Tag sequence:His-tag
- Gene ID:Q8R016
- Protein/peptide name:BLMH
- Sequence:1a.a.-455a.a.