Recombinant Human SIRPG (from Baculovirus), hIgG-His-tag
Catalog # ABNOP8110.100UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:100 µG
- Protein/peptide type:Recombinant
- Source:Baculovirus
- Species:Human
- Tag sequence:hIgG-His-tag
- Gene ID:Q9P1W8
- Protein/peptide name:SIRPG
- Sequence:29a.a.-360a.a.