Recombinant Human CD28 (from Baculovirus), hIgG-His-tag
Catalog # ABNOP8020.100UG
Proveedor: Abnova
Número CAS:
Especificaciones
- Pk:100 µG
- Protein/peptide type:Recombinant
- Source:Baculovirus
- Species:Human
- Tag sequence:hIgG-His-tag
- Gene ID:P10747
- Protein/peptide name:CD28
- Sequence:19a.a.-152a.a.