Order Entry
Spain
ContactUsLinkComponent
 
Recombinant Human GSTM5 (from E. coli), His-tag
  ABNOP7942.100UG
 :  Abnova
 :  
Recombinant Human GSTM5 (from E. coli), His-tag
  ABNOP7942.100UG
 :  Abnova
 :  

 

  • Pk:
    100 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    His-tag
  • Gene ID:
    P46439
  • Protein/peptide name:
    GSTM5
  • Sequence:
    1a.a.-218a.a.