Order Entry
Spain
ContactUsLinkComponent
Recombinant Human ENTPD3 (from E. coli), His-tag
Recombinant Human ENTPD3 (from E. coli), His-tag
Catalog # ABNOP7790.50UG
Supplier:  Abnova
CAS Number:  
Recombinant Human ENTPD3 (from E. coli), His-tag
Catalog # ABNOP7790.50UG
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    His-tag
  • Gene ID:
    NP_001239.2
  • Protein/peptide name:
    ENTPD3
  • Sequence:
    44a.a.-485a.a

Specifications

About this item