Recombinant Mouse TIMP1 (from Baculovirus), His-tag
ABNOP7870.500UG
: Abnova
:
- Pk:500 µG
- Protein/peptide type:Recombinant
- Source:Baculovirus
- Species:Mouse
- Tag sequence:His-tag
- Gene ID:P12032
- Protein/peptide name:TIMP1
- Sequence:25a.a.-205a.a.