Order Entry
Spain
ContactUsLinkComponent
Recombinant Human RAB3IL1 (from E. coli), His-tag
Recombinant Human RAB3IL1 (from E. coli), His-tag
Catalog # ABNOP7758.250UG
Supplier:  Abnova
CAS Number:  
Recombinant Human RAB3IL1 (from E. coli), His-tag
Catalog # ABNOP7758.250UG
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    250 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    His-tag
  • Gene ID:
    NP_037533
  • Protein/peptide name:
    RAB3IL1
  • Sequence:
    1a.a.-382a.a

Specifications

About this item