Recombinant Mouse CD24A (from CHO cells), hFc tag
Catalog # ABNOP7645.50UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:CHO cells
- Species:Mouse
- Tag sequence:hFc tag
- Gene ID:P24807
- Protein/peptide name:CD24A
- Sequence:27a.a.-53a.a.