Recombinant Human GMFB (from E. coli), His-tag
Catalog # ABNOP7625.1MG
Supplier: Abnova
CAS Number:
Specifications
- Pk:1 mg
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:His-tag
- Gene ID:P60983
- Protein/peptide name:GMFB
- Sequence:1a.a.-142a.a.