Skip to Header
Skip to Main Content
Skip to Footer
Avanzado
Avanzado
SDS
Certificados
Datos del fabricante
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Spain
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Recombinant Human SERPINB5 (from E. coli)
Recombinant Human SERPINB5 (from E. coli)
Recombinant Human SERPINB5 (from E. coli)
ABNOP7632.1MG
:
Abnova
:
Recombinant Human SERPINB5 (from E. coli)
ABNOP7632.1MG
:
Abnova
:
Pk:
1 mg
Protein/peptide type:
Recombinant
Source:
E. coli
Species:
Human
Gene ID:
P36952-2
Protein/peptide name:
SERPINB5
Sequence:
1a.a.-231a.a.