Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Spain
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Recombinant Rat OSM (from E. coli)
Recombinant Rat OSM (from E. coli)
Recombinant Rat OSM (from E. coli)
Catalog # ABNOP7637.1MG
Supplier:
Abnova
CAS Number:
Expand image
Recombinant Rat OSM (from E. coli)
Catalog # ABNOP7637.1MG
Supplier:
Abnova
CAS Number:
Specifications
Pk:
1 mg
Protein/peptide type:
Recombinant
Source:
E. coli
Species:
Rat
Gene ID:
Q65Z15
Protein/peptide name:
OSM
Sequence:
26a.a.-239a.a.
Specifications
About this item
Product Details
Product Details