Order Entry
Spain
ContactUsLinkComponent
Recombinant Human YY1 (from E. coli), His-tag
Recombinant Human YY1 (from E. coli), His-tag
Catalog # ABNOP7706.500UG
Proveedor:  Abnova
Número CAS:  
Recombinant Human YY1 (from E. coli), His-tag
Catalog # ABNOP7706.500UG
Proveedor:  Abnova
Número CAS:  

Especificaciones

  • Pk:
    500 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    His-tag
  • Gene ID:
    NP_003394.1
  • Protein/peptide name:
    YY1
  • Sequence:
    1a.a.-414a.a

Especificaciones

Acerca de este artículo