Recombinant E. coli IDNK (from E. coli), His-tag
Catalog # ABNOP7684.500UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:500 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:E. coli
- Tag sequence:His-tag
- Gene ID:NP_418689
- Protein/peptide name:IDNK
- Sequence:1a.a.-187a.a