Recombinant Human SFTPD (from HEK293 cells)
Catalog # ABNOP7538.50UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Gene ID:P35247
- Protein/peptide name:SFTPD
- Sequence:21a.a.-375a.a.