Order Entry
Spain
ContactUsLinkComponent
Recombinant Mouse CXCL9 (from E. coli)
Recombinant Mouse CXCL9 (from E. coli)
Catalog # ABNOP7526.5UG
Supplier:  Abnova
CAS Number:  
Recombinant Mouse CXCL9 (from E. coli)
Catalog # ABNOP7526.5UG
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    5 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Gene ID:
    P18340
  • Protein/peptide name:
    CXCL9
  • Sequence:
    22a.a.-126a.a.

Specifications

About this item