Recombinant Human FGF21 (from CHO cells), His-tag
Catalog # ABNOP7443.10UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Source:CHO cells
- Species:Human
- Tag sequence:His-tag
- Gene ID:Q9NSA1
- Protein/peptide name:FGF21
- Sequence:29a.a.-209a.a.