Recombinant Mouse IL12A & IL12B (from CHO cells)
Catalog # ABNOP7447.10UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Source:CHO cells
- Species:Mouse
- Gene ID:P43432
- Protein/peptide name:IL12A & IL12B
- Sequence:23a.a.-335a.a.