Order Entry
Spain
ContactUsLinkComponent
Recombinant Mouse FLT3L (from CHO cells), His-tag
Recombinant Mouse FLT3L (from CHO cells), His-tag
Catalog # ABNOP7453.10UG
Supplier:  Abnova
CAS Number:  
Recombinant Mouse FLT3L (from CHO cells), His-tag
Catalog # ABNOP7453.10UG
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Mouse
  • Tag sequence:
    His-tag
  • Gene ID:
    P49772
  • Protein/peptide name:
    FLT3L
  • Sequence:
    27a.a.-188a.a.

Specifications

About this item