Order Entry
Spain
ContactUsLinkComponent
Recombinant Human IGF1 (from E. coli)
Recombinant Human IGF1 (from E. coli)
  ABNOP7326.10UG
 :  Abnova
 :  
Recombinant Human IGF1 (from E. coli)
  ABNOP7326.10UG
 :  Abnova
 :  

 

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Gene ID:
    P05019
  • Protein/peptide name:
    IGF1
  • Sequence:
    49a.a.-118a.a