Order Entry
Spain
ContactUsLinkComponent
Recombinant Mouse FGF1 (from E. coli)
Recombinant Mouse FGF1 (from E. coli)
Catalog # ABNOP7336.10UG
Supplier:  Abnova
CAS Number:  
Recombinant Mouse FGF1 (from E. coli)
Catalog # ABNOP7336.10UG
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Gene ID:
    P61148
  • Protein/peptide name:
    FGF1
  • Sequence:
    16a.a.-155a.a.

Specifications

About this item