Recombinant Human OSM (from E. coli)
Catalog # ABNOP7340.10UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Gene ID:P13725
- Protein/peptide name:OSM
- Sequence:26a.a.-252a.a