Recombinant Human IL2 (from HEK293 cells)
Catalog # ABNOP7343.10UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Gene ID:P60568
- Protein/peptide name:IL2
- Sequence:21a.a.-153a.a