Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Spain
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Antibodies
Anti-PHD1 Rabbit Recombinant Antibody
Anti-PHD1 Rabbit Recombinant Antibody
Anti-PHD1 Rabbit Recombinant Antibody
Catalog # ABNORAB02396.100UL
Supplier:
Abnova
Expand image
Anti-PHD1 Rabbit Recombinant Antibody
Catalog # ABNORAB02396.100UL
Supplier:
Abnova
Specifications
Antigen symbol:
PHD1
Clonality:
Recombinant
Host:
Rabbit
Immunogen:
Rabbit monoclonal antibody raised against recombinant PHD1.
Pk:
100 µl
Specifications
About this item
Product Details
Product Details