Anti-WASL Rabbit Recombinant Antibody [clone: R02-7K5]
Catalog # ABNORAB02042.100UL
Supplier: Abnova
Specifications
- Antigen symbol:WASL
- Clonality:Recombinant
- Clone:R02-7K5
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human WASL.
- Pk:100 µl