Order Entry
Spain
ContactUsLinkComponent
Anti-H2AZ1 Rabbit Recombinant Antibody [clone: R09-3H9]
Anti-H2AZ1 Rabbit Recombinant Antibody [clone: R09-3H9]
Catalog # ABNORAB02225.100UL
Supplier:  Abnova
Anti-H2AZ1 Rabbit Recombinant Antibody [clone: R09-3H9]
Catalog # ABNORAB02225.100UL
Supplier:  Abnova

Specifications

  • Antigen symbol:
    H2AZ1
  • Clonality:
    Recombinant
  • Clone:
    R09-3H9
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human H2AZ1.
  • Pk:
    100 µl

Specifications

About this item