Order Entry
Spain
ContactUsLinkComponent
Anti-GYS1 Rabbit Recombinant Antibody [clone: R03-3A1]
Anti-GYS1 Rabbit Recombinant Antibody [clone: R03-3A1]
Catalog # ABNORAB02244.100UL
Supplier:  Abnova
Anti-GYS1 Rabbit Recombinant Antibody [clone: R03-3A1]
Catalog # ABNORAB02244.100UL
Supplier:  Abnova

Specifications

  • Antigen symbol:
    GYS1
  • Clonality:
    Recombinant
  • Clone:
    R03-3A1
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human GYS1.
  • Pk:
    100 µl

Specifications

About this item