Anti-SMN1 Rabbit Recombinant Antibody [clone: R05-3B5]
Catalog # ABNORAB02258.100UL
Proveedor: Abnova
Especificaciones
- Antigen symbol:SMN1
- Clonality:Recombinant
- Clone:R05-3B5
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human SMN1.
- Pk:100 µl