Anti-H3C1 Rabbit Recombinant Antibody [clone: R03-7K5]
ABNORAB02222.100UL
: Abnova
- Antigen symbol:H3C1
- Clonality:Recombinant
- Clone:R03-7K5
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human H3C1.
- Pk:100 µl