Anti-H2BC12 Rabbit Recombinant Antibody [clone: R07-8K4]
Catalog # ABNORAB02223.100UL
Proveedor: Abnova
Especificaciones
- Antigen symbol:H2BC12
- Clonality:Recombinant
- Clone:R07-8K4
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human H2BC12.
- Pk:100 µl