Anti-H2AX Rabbit Recombinant Antibody [clone: R09-4A7]
Catalog # ABNORAB02266.100UL
Supplier: Abnova
Specifications
- Antigen symbol:H2AX
- Clonality:Recombinant
- Clone:R09-4A7
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human H2AX.
- Pk:100 µl