Order Entry
Spain
ContactUsLinkComponent
Anti-MTDH Rabbit Recombinant Antibody [clone: R05-5G3]
Anti-MTDH Rabbit Recombinant Antibody [clone: R05-5G3]
  ABNORAB01416.100UL
 :  Abnova
Anti-MTDH Rabbit Recombinant Antibody [clone: R05-5G3]
  ABNORAB01416.100UL
 :  Abnova

 

  • Antigen symbol:
    MTDH
  • Clonality:
    Recombinant
  • Clone:
    R05-5G3
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human MTDH.
  • Pk:
    100 µl