Anti-ICAM1 Rabbit Recombinant Antibody [clone: R09-7A9]
Catalog # ABNORAB02190.100UL
Supplier: Abnova
Specifications
- Antigen symbol:ICAM1
- Clonality:Recombinant
- Clone:R09-7A9
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human ICAM1.
- Pk:100 µl