Anti-HSF1 Rabbit Recombinant Antibody [clone: R07-0G0]
ABNORAB02201.100UL
: Abnova
- Antigen symbol:HSF1
- Clonality:Recombinant
- Clone:R07-0G0
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human HSF1.
- Pk:100 µl