Anti-STING1 Rabbit Recombinant Antibody [clone: R06-9A7]
Catalog # ABNORAB01862.100UL
Supplier: Abnova
Specifications
- Simbolo antígeno:STING1
- Clonalidad:Recombinant
- Clon:R06-9A7
- Huésped:Rabbit
- Reactividad:Human
- Inmunógeno:Recombinant rabbit monoclonal antibody raised against human STING1.
- Env:100 µl