Order Entry
Spain
ContactUsLinkComponent
Anti-CD39 Rabbit Recombinant Antibody [clone: R09-3I2]
Anti-CD39 Rabbit Recombinant Antibody [clone: R09-3I2]
Catalog # ABNORAB01523.100UL
Supplier:  Abnova
Anti-CD39 Rabbit Recombinant Antibody [clone: R09-3I2]
Catalog # ABNORAB01523.100UL
Supplier:  Abnova

Specifications

  • Antigen symbol:
    CD39
  • Clonality:
    Recombinant
  • Clone:
    R09-3I2
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against synthetic peptide of human CD39.
  • Pk:
    100 µl

Specifications

About this item