Order Entry
Spain
ContactUsLinkComponent
 
Anti-Caspase-9 Rabbit Recombinant Antibody [clone: R04-8I4]
  ABNORAB01536.100UL
 :  Abnova
Anti-Caspase-9 Rabbit Recombinant Antibody [clone: R04-8I4]
  ABNORAB01536.100UL
 :  Abnova

 

  • Antigen symbol:
    Caspase-9
  • Clonality:
    Recombinant
  • Clone:
    R04-8I4
  • Host:
    Rabbit
  • Reactivity:
    Mouse
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against mouse Caspase-9.
  • Pk:
    100 µl