Order Entry
Spain
ContactUsLinkComponent
 
Anti-Casein Kinase 1 alpha Rabbit Recombinant Antibody [clone: R05-6C6]
  ABNORAB01540.100UL
 :  Abnova
Anti-Casein Kinase 1 alpha Rabbit Recombinant Antibody [clone: R05-6C6]
  ABNORAB01540.100UL
 :  Abnova

 

  • Antigen symbol:
    Casein Kinase 1 alpha
  • Clonality:
    Recombinant
  • Clone:
    R05-6C6
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against synthetic peptide of human Casein Kinase 1 alpha.
  • Pk:
    100 µl