Anti-CaMKII delta Rabbit Recombinant Antibody [clone: R03-8A9]
ABNORAB01543.100UL
: Abnova
- Antigen symbol:CaMKII delta
- Clonality:Recombinant
- Clone:R03-8A9
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against synthetic peptide of human CaMKII delta.
- Pk:100 µl