Anti-C3 Rabbit Recombinant Antibody [clone: R06-2C2]
Catalog # ABNORAB01548.100UL
Supplier: Abnova
Specifications
- Antigen symbol:C3
- Clonality:Recombinant
- Clone:R06-2C2
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against synthetic peptide of human C3.
- Pk:100 µl