Anti-BMP4 Rabbit Recombinant Antibody [clone: R01-2G4]
Catalog # ABNORAB01553.100UL
Supplier: Abnova
Specifications
- Antigen symbol:BMP4
- Clonality:Recombinant
- Clone:R01-2G4
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against synthetic peptide of human BMP4.
- Pk:100 µl