Order Entry
Spain
ContactUsLinkComponent
Anti-beta Catenin Rabbit Recombinant Antibody [clone: R07-9D5]
Anti-beta Catenin Rabbit Recombinant Antibody [clone: R07-9D5]
Catalog # ABNORAB01560.100UL
Supplier:  Abnova
Anti-beta Catenin Rabbit Recombinant Antibody [clone: R07-9D5]
Catalog # ABNORAB01560.100UL
Supplier:  Abnova

Specifications

  • Antigen symbol:
    beta Catenin
  • Clonality:
    Recombinant
  • Clone:
    R07-9D5
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against synthetic peptide of human beta Catenin.
  • Pk:
    100 µl

Specifications

About this item