Anti-CD57 Mouse Monoclonal Antibody [clone: HCD57] (FITC)
Catalog # ABNOMAB23620.500UG
Supplier: Abnova
Specifications
- Antigen symbol:CD57
- Clonality:Monoclonal
- Clone:HCD57
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Mouse
- Reactivity:Human
- Immunogen:Mouse FITC monoclonal antibody raised against synthetic peptide of human CD57.
- Pk:500 µG